Order Entry
Poland
ContactUsLinkComponent
Barium chloride 10% in aqueous solution
Irritant
Barium chloride 10% in aqueous solution
Catalog # 164735P
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Barium chloride 10% in aqueous solution
Catalog # 164735P
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay (W/V):
    9.95 - 10.00 %
  • pH (25°C):
    6.5 - 7.5

Specification Test Results

About this item

 :
 :208.24 g/mol
Boiling Pt:>100 °C (1013 hPa)
Storage Temperature:Ambient
EINECS:233-788-1
UN:3287
ADR:6.1,III
 :GHS07
CAS Number:10361-37-2
Formula:BaCl₂
MDL Number:MFCD00003445