Order Entry
Poland
ContactUsLinkComponent
Potassium iodide 40 g/l in aqueous solution, GPR RECTAPUR®
Health
Potassium iodide 40 g/l in aqueous solution, GPR RECTAPUR®
Catalog # 26852.293
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Potassium iodide 40 g/l in aqueous solution, GPR RECTAPUR®
Catalog # 26852.293
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay:
    Min. 40 g/l

Specification Test Results

About this item

 :
 :166 g/mol
EINECS:231-659-4
 :GHS08
Storage Temperature:Ambient
CAS Number:7681-11-0
Formula:Kl
MDL Number:MFCD00011405