Hydrogen peroxide 3% (w/w) in water stabilised, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7722-84-1
:
Formula:H₂O₂
MDL Number:MFCD00011333
Boiling Pt:100 °C (1013 hPa)
Density:1.00 g/cm³
:Stabilised with acetanilide ~200 ppm
:34.01 g/mol
EINECS:231-765-0
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance (colorless liquid):complying
- Assay:3.0 - 4.0 Gew.%
- pH (direct, 20 °C):4 - 7
- Stabilizer (Acetanilide):150 - 250 ppm
- Contains:~200 ppm acetanilide as stabilizer
Specification Test Results
Frequently Bought Together