Order Entry
Poland
ContactUsLinkComponent
Acrylamide 40% in aqueous solution, Acryl 40™, Proteomics Grade
Toxic
Health
Irritant
Acrylamide 40% in aqueous solution, Acryl 40™, Proteomics Grade
Catalog # M121-500ML
Supplier:  VWR Chemicals
CAS Number:  
undefined
Toxic
Health
Irritant
Acrylamide 40% in aqueous solution, Acryl 40™, Proteomics Grade
Catalog # M121-500ML
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Acrylic Acid:
    <0.005%
  • Conductivity:
    <5 umhos
  • DNase:
    none detected
  • Electrophoresis:
    Pass
  • pH @25 °C:
    6 - 7
  • Protease:
    none detected
  • RNase:
    none detected

Specification Test Results

About this item

 :
 :71.08 g/mol
Storage Temperature:Refrigerator
EINECS:201-173-7
UN:3426
ADR:6.1,III
 :GHS06
 :GHS08
 :GHS07
CAS Number:79-06-1
Formula:C₃H₅NO
MDL Number:MFCD00008032