Order Entry
Poland
ContactUsLinkComponent
(±)-2-Methyl-2,4-pentanediol, Sigma-Aldrich®
(±)-2-Methyl-2,4-pentanediol, Sigma-Aldrich®
Catalog # SIALM9671-50G
Supplier:  Merck
CAS Number:  
(±)-2-Methyl-2,4-pentanediol, Sigma-Aldrich®
Catalog # SIALM9671-50G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :118.18 g/mol
Merck Index:13,04730
Boiling Pt:198 °C (1013 hPa)
Melting Pt:–40 °C
Density:0.923 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
Flash Pt:93 °C
Storage Temperature:Ambient
EINECS:203-489-0
 :GHS07
CAS Number:107-41-5
Formula:C₆H₁₄O₂
MDL Number:MFCD00004547