Order Entry
Poland
ContactUsLinkComponent
 
 
 
 
Benzyl chloride 99.5+% stabilised, extra pure
 
undefined
 
 
 
Benzyl chloride 99.5+% stabilised, extra pure

 

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5370 to 1.5390 (20°C, 589 nm)
  • GC:
    ≥99.50 %
  • Color scale:
    ≤20 APHA
  • Stabilizer:
    0.25 % propylene oxide

 

 

Frequently Bought Together