Order Entry
Poland
ContactUsLinkComponent
 
Health
Irritant
Dichloromethane, dehydrated (max. 0.01% H₂O) ≥99% stabilised, GPR RECTAPUR® for synthesis
Catalog # 23349.363
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Irritant
Dichloromethane, dehydrated (max. 0.01% H₂O) ≥99% stabilised, GPR RECTAPUR® for synthesis
Catalog # 23349.363
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle, fluorinated
  • Pk:
    5 L
  • Seq:
    5190

Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Evaporation residue:
    Max. 50 ppm
  • Water:
    Max. 100 ppm

Specification Test Results

About this item

 :
 :84.93 g/mol
Merck Index:12,06140
Boiling Pt:39.8 °C (1013 hPa)
Melting Pt:–95 °C
Density:1.322 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-838-9
UN:1593
ADR:6.1,III
 :Stabilised with ethanol 0.1 %
REACH:01-2119480404-41
 :GHS08
 :GHS07
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881