L(+)-Cysteine hydrochloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:52-89-1
:
Formula:HSCH₂CH(NH₂)COOH·HCl
MDL Number:MFCD00064553
:157.62 g/mol
Merck Index:13,02810
Melting Pt:175 °C
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:200-157-7
Frequently Bought Together