Polyvinylpyrrolidone, M.W. ∼ 55,000, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP
CAS Number:9003-39-8
:
Formula:(C₆H₉NO)n
MDL Number:MFCD01076626
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together