Order Entry
Poland
ContactUsLinkComponent
Polyvinylpyrrolidone (K30), BioChemica®
Polyvinylpyrrolidone (K30), BioChemica®
Supplier:  PanReac AppliChem
Certificates
Polyvinylpyrrolidone (K30), BioChemica®
Supplier:  PanReac AppliChem
Certificates

Specification Test Results

  • Assay (from N):
    12.0 - 12.8 %
  • Solubility (10 %; H2O):
    clear, colorless to yellowish
  • pH (5 %; H2O):
    3.0 - 5.0
  • Heavy metals (as Pb):
    max. 0.001 %
  • Water (K.F.):
    max. 5 %

Specification Test Results

Related Information

Frequently Bought Together