Tween® 60 (Polisorbat), Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
Synonyms: Polyoxyethylene sorbitan monostearate
- CAS Number:9005-67-8
- Formula:C₆₄H₁₂₆O₂₆
- MDL Number:MFCD01780110
- Boiling Pt:∼110 °C (1013 hPa)
- Density:1,08 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ100
- Flash Pt:>149 °C
- Storage Temperature:Temperatura otoczenia
- EINECS:500-020-4
3 Options Available Below
Product Details & Documents
Synonyms: Polyoxyethylene sorbitan monostearate
- CAS Number:9005-67-8
- Formula:C₆₄H₁₂₆O₂₆
- MDL Number:MFCD01780110
- Boiling Pt:∼110 °C (1013 hPa)
- Density:1,08 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ100
- Flash Pt:>149 °C
- Storage Temperature:Temperatura otoczenia
- EINECS:500-020-4



