Order Entry
Poland
Orders LinkContactUsLinkComponent
Tween® 20 (Polysorbate), Millipore®

Tween® 20 (Polysorbate), Millipore®

Glass bottle, 100 mL
Catalog # 655204-100
Tween® 20 (Polysorbate), Millipore®
Catalog # 655204-100
CAS Number:  9005-64-5
Tween® 20 (Polysorbate), Millipore®

Glass bottle, 100 mL

CAS Number:  9005-64-5
Price
299 zł
Log in for availability
1 * 100 mL
Product Details & Documents
Synonyms:  Polyoxyethylene sorbitan monolaurate
  • CAS Number:9005-64-5
  • Formula:C₅₈H₁₁₄O₂₆
  • MDL Number:MFCD00165986
  • Molecular Weight:1227.53 g/mol
  • Density:1.1 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:500-018-3
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.