Order Entry
Poland
ContactUsLinkComponent
L(+)-Asparagine monohydrate ≥99.0% (dry basis) for biochemistry pH 4.0 - 5.5 (20 g/l, H₂O, 20 °C), Millipore®
L(+)-Asparagine monohydrate ≥99.0% (dry basis) for biochemistry pH 4.0 - 5.5 (20 g/l, H₂O, 20 °C), Millipore®
Supplier:  Merck
Certificates
L(+)-Asparagine monohydrate ≥99.0% (dry basis) for biochemistry pH 4.0 - 5.5 (20 g/l, H₂O, 20 °C), Millipore®
Supplier:  Merck

Specification Test Results

  • Assay (perchloric acid titration, calculated on dried substance):
    ≥ 99.0 %
  • Identity (IR-spectrum):
    passes test
  • Appearance:
    white to almost white crystalline powder or colorless crystals
  • Appearance of solution (20 g/l, CO₂-free water):
    clear and colorless
  • Spec. rotation (α 20/D, 100 g/l, hydrochloric acid 3 mol/l, calc. on dried substance):
    +33.7 to +36.0
  • Chloride (Cl):
    ≤ 0.02 %
  • Sulphate (SO₄):
    ≤ 0.02 %
  • Heavy metals (as Pb):
    ≤ 0.001 %
  • As (Arsenic):
    ≤ 0.0005 %
  • Ca (Calcium):
    ≤ 0.001 %
  • Co (Cobalt):
    ≤ 0.0005 %
  • Fe (Iron):
    ≤ 0.0005 %
  • K (Potassium):
    ≤ 0.0005 %
  • Mg (Magnesium):
    ≤ 0.0005 %
  • Na (Sodium):
    ≤ 0.005 %
  • NH4 (Ammonium):
    ≤ 0.01 %
  • Zn (Zinc):
    ≤ 0.0005 %
  • Aspartic acid:
    ≤ 0.5 %
  • other foreign amino acids:
    ≤ 0.3 %
  • Other ninhydrine positive substances:
    ≤ 0.1 %
  • Loss on drying (130 °C, 3 h):
    10.5 - 12.5 %

Specification Test Results

Related Information

Frequently Bought Together