Order Entry
Poland
ContactUsLinkComponent
Polyethylene glycol 4,000 for synthesis, Sigma-Aldrich®
Polyethylene glycol 4,000 for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Polyethylene glycol 4,000 for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    500 g/l (20 °C)
  • Melting Point:
    53 - 58 °C
  • Molar Mass:
    3500 - 4500 g/mol
  • Bulk Density:
    400 - 500 kg/m3
  • Vapor Pressure:
     - 0.01 hPa (20 °C)
  • Flash Point:
    250 °C
  • Density:
    1.2 g/cm3 (20 °C)
  • pH:
    5 - 7 (100 g/l, H2O, 20 °C)
  • Ignition Point:
    320 °C DIN 51794
  • Viscosity kinematic:
    76 - 110 mm2/s (98 °C)

Specification Test Results

Related Information

Frequently Bought Together