Order Entry
Poland
ContactUsLinkComponent
 
Polydimethylsiloxane, 30000 cSt, Sigma-Aldrich®
Catalog # SIAL378429-250ML
Supplier:  Merck
CAS Number:  
Polydimethylsiloxane, 30000 cSt, Sigma-Aldrich®
Catalog # SIAL378429-250ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

About this item

 :
CAS Number:63148-62-9
 :9016-00-6
Formula:(CH₃)₃SiO(Si(CH₃)₂O)n
MDL Number:MFCD00132673