Order Entry
Poland
ContactUsLinkComponent
Poly(dimethylsiloxane-co-methylphenylsiloxane), 125 cSt (25°C), Sigma-Aldrich®
Poly(dimethylsiloxane-co-methylphenylsiloxane), 125 cSt (25°C), Sigma-Aldrich®
Catalog # SIAL378488-50ML
Supplier:  Merck
CAS Number:  
Poly(dimethylsiloxane-co-methylphenylsiloxane), 125 cSt (25°C), Sigma-Aldrich®
Catalog # SIAL378488-50ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

About this item

 :
Boiling Pt:∼35 °C (1013 hPa)
Density:0.982 g/cm³ (25 °C)
 :M-Clarity™ quality level = MQ100
Flash Pt:>101.1 °C (closed cup)
Storage Temperature:Ambient
CAS Number:63148-52-7