Starch, soluble ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9005-84-9
:
Formula:(C₆H₁₀O₅)n
MDL Number:MFCD00082026
:162.14 g/mol
Merck Index:13,08878
Melting Pt:256…258 °C
Storage Temperature:Ambient
EINECS:232-686-4
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Solubility:Passes test
- Residue after ignition:≤0.4 %
- pH:5.0 to 7.0 (2 % solution at 25.0°C)
- Sensitivity:Passes test
Specification Test Results
Frequently Bought Together