Starch (from potato), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:9005-25-8
:
Formula:(C₆H₁₀O₅)n
MDL Number:MFCD00082026
Molecular Weight:162.14 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:232-679-6
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together