Agar, Millipore®
Glass bottle for solids, 500 g
Catalog # SIAL05038-500G
Supplier: Merck
Jump to:
Product Details & Documents
Synonyms: Agar Agar
- CAS Number:9002-18-0
- Formula:C₁₂nH₁₈nOn₉n
- MDL Number:MFCD00081288
- Merck Index:13,00184
- Melting Pt:90 °C
- :M-Clarity™ quality level = MQ100
- Storage Temperature:Ambient
- EINECS:232-658-1
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Cat. No.:SIAL05038-500G



