Order Entry
Poland
Orders LinkContactUsLinkComponent
Gentamicin sulphate

Gentamicin sulphate

Gentamicin sulphate
Supplier:  PanReac AppliChem
Certificates
Gentamicin sulphate

2 Options Available Below

Product Details & Documents
Synonyms:  NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C
  • CAS Number:1405-41-0
  • MDL Number:MFCD00270181
  • Merck Index:13,04403
  • Melting Pt:218…237 °C
  • Storage Temperature:Ambient
  • EINECS:215-778-9

Documents

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 201 zł
    201 zł
    1 * 5 g
       201 zł

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 529 zł
    529 zł
    1 * 25 g
       529 zł