Order Entry
Poland
ContactUsLinkComponent
cis-Platin for synthesis, Sigma-Aldrich®
Toxic
Health
cis-Platin for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Toxic
Health
cis-Platin for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    2.5 g/l (20 °C)
  • Molar Mass:
    300.05 g/mol
  • Bulk Density:
    850 kg/m3
  • Density:
    3.7 g/cm3 (20 °C)

Specification Test Results

Related Information

Frequently Bought Together