


cis-Platin ≥64.5%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15663-27-1
:
Formula:H₆Cl₂N₂Pt
MDL Number:MFCD00011623
:300.05 g/mol
Merck Index:14,2317
Melting Pt:270 °C (dec.)
Density:3.738 g/cm³ (20 °C)
EINECS:239-733-8
UN:3288
ADR:6.1,II
:GHS06
:GHS08
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together