Złóż zamówienie
Poland
ContactUsLinkComponent
 
 
 
 
cis-Platyna 99.99% (trace metals basis)
 
undefined
 
 
 
cis-Platyna 99.99% (trace metals basis)

 

  • Appearance:
    Yellow to orange Powder or crystals
  • Infrared spectrum:
    Conforms
  • Assay:
    64.0 to 66.0 % (Pt)
  • Total trace metal impurities:
    ≤0.01 %

 

 

Frequently Bought Together