



Boron trifluoride diethyl ether complex for synthesis, Sigma-Aldrich®
: Merck
:109-63-7
:
:C₄H₁₀BF₃O
:MFCD00013194
:141.93 g/mol
:13,01340
:126 °C (1013 hPa)
:–60 °C
:1.125 g/cm³ (25 °C)
:Ambient
:203-689-8
:2604
:8 (3),I
:M-Clarity™ quality level = MQ200
:59 °C
:GHS02
:GHS05
:GHS08
:GHS07
- Density:1.013 g/cm3 (20 °C)
- pH:0.5 (H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Frequently Bought Together