Order Entry
Poland
ContactUsLinkComponent
 
Trifluoroacetic acid-[D₁] 99.5 atom % D for NMR spectroscopy
  ACRO325330100
 :  
undefined
Trifluoroacetic acid-[D₁] 99.5 atom % D for NMR spectroscopy
  ACRO325330100
 :  

 

  • Pack type:
    Ampoule
  • Pk:
    10 mL
  • Seq:
    2076
  • Pk Info:
    10 x 1 ml

 

 

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.05 % (Coulometric)
  • D-Enrichment:
    ≥99.4 %

 

 

 :
 :115.02 g/mol
 :75 °C (1013 hPa)
 :–15 °C
 :1.5 g/cm³
 :209-961-2
 :2699
 :8,I
 :GHS07
 :GHS05
 :599-00-8
 :F₃CCOOD
 :MFCD00044563