



Petroleum spirit, 40…60 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:64742-49-0
:64742-82-1
MDL Number:MFCD00081849
Boiling Pt:40…60 °C (1013 hPa)
Density:0.645…0.665 g/cm³ (20 °C)
Flash Pt:–40 °C
Storage Temperature:Ambient
EINECS:265-151-9
UN:1268
ADR:3,II
REACH:01-2119474679-18
:GHS02
:GHS08
:GHS07
:GHS09
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Acidity or alkalinity:Max. 0.0002 meq/g
- Bromine index:Max. 1
- boiling range (40-60 °C):75% min.
- Substances coloured by H2SO4:Max. 150 APHA
- Benzene:Max. 0.15 %
- Evaporation residue:Max. 10 ppm
- Total S (as SO4):Max. 50 ppm
- Water:Max. 0.015 %
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 30L Metal drum for liquids UN X (ZPACK12124125) 5L Tin bottle for liquids UN Y (ZPACK12321007)
Packaging Information
23826_PETROLEUM SPIRIT 40-60°C _BSE_REV0
BSE/TSE Certificates
spec_VWRC23826.293 spec_VWRC23826.293 spec_VWRC23826.327 spec_VWRC23826.360 spec_VWRC23826.464
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Frequently Bought Together