
Soda lime ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:8006-28-8
:
MDL Number:MFCD00134094
Merck Index:14,8569
Storage Temperature:Ambient
UN:1907
ADR:8,III
:GHS05
Related Information
Frequently Bought Together