Order Entry
Poland
ContactUsLinkComponent
 
Platinum, powder, Sigma-Aldrich®
Catalog # SIAL204013-1G
Supplier:  Merck
CAS Number:  
Platinum, powder, Sigma-Aldrich®
Catalog # SIAL204013-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :195.08 g/mol
Boiling Pt:4530 °C (1013 hPa)
Melting Pt:1773 °C
Density:21.4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-116-1
UN:3089
ADR:4.1,II
 :M-Clarity™ quality level = MQ100
CAS Number:7440-06-4
Formula:Pt
MDL Number:MFCD00011179