Platinum, Sigma-Aldrich®
SIAL349399-3.8G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:3,8 g
- Seq:3152
:
:195.08 g/mol
:12,07689
:3827 °C (1013 hPa)
:1769 °C
:21.447 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:Ambient
:231-116-1
:7440-06-4
:Pt
:MFCD00011179