Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Platinum
Platinum ≥99.9% (metals basis), wire, Ø 0.01 mm (0.0004 in)
Platinum ≥99.9% (metals basis), wire, Ø 0.01 mm (0.0004 in)
44196.G1
:
Thermo Fisher Scientific
:
Platinum ≥99.9% (metals basis), wire, Ø 0.01 mm (0.0004 in)
44196.G1
:
Thermo Fisher Scientific
:
Pk:
1 m
:
7440-06-4
:
:
Pt
:
MFCD00011179
:
195.08 g/mol
:
12,07689
:
3827 °C (1013 hPa)
:
1769 °C
:
21.447 g/cm³ (20 °C)
:
Ambient
:
231-116-1