Order Entry
Poland
ContactUsLinkComponent
Platinum ≥99.9% (metals basis), foil 2.0 mm (0.08 in) thick
Platinum ≥99.9% (metals basis), foil 2.0 mm (0.08 in) thick
Catalog # 12614.FF
CAS Number:  
Platinum ≥99.9% (metals basis), foil 2.0 mm (0.08 in) thick
Catalog # 12614.FF
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    1643
  • Pk Info:
    25×25 mm

Specifications

About this item

 :
 :195.08 g/mol
Merck Index:12,07689
Boiling Pt:3827 °C (1013 hPa)
Melting Pt:1769 °C
Density:21.447 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-116-1
CAS Number:7440-06-4
Formula:Pt
MDL Number:MFCD00011179