Nickel, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7440-02-0
:
Formula:Ni
MDL Number:MFCD00011137
:58.69 g/mol
Merck Index:12,06582
Boiling Pt:3007 °C (1013 hPa)
Melting Pt:1453 °C
Density:8.9 g/cm³ (20 °C)
UN:3089
ADR:4.1,III
:M-Clarity™ quality level = MQ100
EINECS:231-111-4
:GHS08
Storage Temperature:Ambient
Frequently Bought Together