Iridium ≥99.7% (metals basis), foil, Thickness: 1.0 mm (0.04 in)
Catalog # 11525.FF
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:1643
- Pk Info:25×25 mm
Specifications
About this item
:
:192.22 g/mol
Boiling Pt:5300 °C (1013 hPa)
Melting Pt:2450 °C
Storage Temperature:Ambient
EINECS:231-095-9
CAS Number:7439-88-5
Formula:Ir
MDL Number:MFCD00011062