0 zł
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Poland
ContactUsLinkComponent
Tungsten
Tungsten, gauze woven from wire 20 mesh, Ø 0.127 mm (0.005 in)
Tungsten, gauze woven from wire 20 mesh, Ø 0.127 mm (0.005 in)
:
Thermo Fisher Scientific
Tungsten, gauze woven from wire 20 mesh, Ø 0.127 mm (0.005 in)
:
Thermo Fisher Scientific
:
7440-33-7
:
:
W
:
MFCD00011461
:
183.84 g/mol
:
12,09945
:
5660 °C (1013 hPa)
:
3410 °C
:
19 g/cm³ (20 °C)
:
Ambient
:
231-143-9
msds_ALFA040922
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
N,N-Dimethylformamide, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Dimethyl sulphoxide ≥99.7% (by GC), LiChrosolv® for liquid chromatography, Supelco®
: Merck
Dimethyl sulphoxide ≥99.99% (by anhydrous basis), PESTINORM® for headspace gas chromatography
: VWR Chemicals
Chloroform 99.0-99.6% stabilised, GPR RECTAPUR®
: VWR Chemicals
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Methanol ≥99.9% (by GC), LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Tetrahydrofuran ≥99.5% (by anhydrous basis) stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
2-Propanol, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Ethyl acetate, EMPLURA®, Supelco®
: Merck
Ethanol ≥99.9% (by GC), LiChrosolv®, gradient grade for liquid chromatography, Supelco®
: Merck
Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
2-Propanol ≥99.5% (by GC), EMPARTA® ACS analytical reagent, Supelco®
: Merck
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals