Order Entry
Poland
ContactUsLinkComponent
 
Flammable
Scandium ≥99.9% (REO, rare earth oxide basis), pieces, distilled dendritic
Catalog # 39996.04
CAS Number:  
Flammable
Scandium ≥99.9% (REO, rare earth oxide basis), pieces, distilled dendritic
Catalog # 39996.04
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2 g
  • Seq:
    3107

Specifications

About this item

 :
 :44.96 g/mol
Merck Index:12,08537
Boiling Pt:2730 °C (1013 hPa)
Melting Pt:1541 °C
Density:2.989 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-129-2
UN:3178
ADR:4.1,III
 :Packaged under argon.
 :GHS02
CAS Number:7440-20-2
Formula:Sc
MDL Number:MFCD00016323