Order Entry
Poland
ContactUsLinkComponent
Erbium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.1 mm (0.004 in)
Flammable
Erbium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.1 mm (0.004 in)
Catalog # 10170.FI
CAS Number:  
Flammable
Erbium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.1 mm (0.004 in)
Catalog # 10170.FI
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    1670
  • Pk Info:
    50×50 mm

Specifications

About this item

 :
 :167.26 g/mol
Storage Temperature:Ambient
EINECS:231-160-1
UN:3178
ADR:4.1,II
 :GHS02
CAS Number:7440-52-0
Formula:Er
MDL Number:MFCD00010987