Silicon ≥99.999% (metals basis), rods, Ø 5.08cm (2.0 in)
Catalog # 39103.DH
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:25 mm
- Seq:1640
Specifications
About this item
:
:28.09 g/mol
Merck Index:12,08635
Boiling Pt:2355 °C (1013 hPa)
Melting Pt:1410 °C
Density:2.34 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-130-8
CAS Number:7440-21-3
Formula:Si
MDL Number:MFCD00085311