Indium ≥99.99% (metals basis), wire, Ø 2.0 mm (0.08 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-74-6
:
Formula:In
MDL Number:MFCD00134048
:114.82 g/mol
Merck Index:13,04971
Boiling Pt:2000 °C (1013 hPa)
Melting Pt:156 °C
Density:7.3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-180-0
Frequently Bought Together