
Gallium 99.99% (trace metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-55-3
:
Formula:Ga
MDL Number:MFCD00134045
:69.72 g/mol
Merck Index:12,04364
Boiling Pt:2403 °C (1013 hPa)
Melting Pt:29.7 °C
Density:5.9 g/cm³
Storage Temperature:Refrigerator
EINECS:231-163-8
UN:2803
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Silver white to silver grey Metallic solid or liquid
- Total trace metal impurities:≤0.01 %
Specification Test Results
Frequently Bought Together