Order Entry
Poland
ContactUsLinkComponent
Aluminium oxide 20% (w/w) in water, nanoparticles 30 - 60 nm (TEM), Sigma-Aldrich®
Aluminium oxide 20% (w/w) in water, nanoparticles 30 - 60 nm (TEM), Sigma-Aldrich®
Catalog # SIAL642991-100ML
Supplier:  Merck
CAS Number:  
Aluminium oxide 20% (w/w) in water, nanoparticles 30 - 60 nm (TEM), Sigma-Aldrich®
Catalog # SIAL642991-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • ICP Major Analysis (Confirms Aluminum Component):
    Confirmed
  • Loading, Disperison:
    19.0 - 21.0% wt.
  • Particle Size (TEM):
    30 - 60 nm

Specification Test Results

About this item

 :
 :101.96 g/mol
Density:1.06 g/ml (25 °C)
 :M-Clarity™ quality level = MQ100
CAS Number:1344-28-1
 :11092-32-3
Formula:Al₂O₃
MDL Number:MFCD00003424