Human Cystatin C
numer katalogowy ENZOBMLSE4790100
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:100 µG
- Species:Human
- Seq:02
- Protein/peptide name:Cystatin C
Specifications
About this item
Protease inhibitor
Isolated from human urine.