Z-VEID-FMK
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Caspase-6 inhibitor
Irreversible, cell permeable inhibitor of caspase-6. The inhibitor is designed as a methyl ester to facilitate cell permeability.
Specifications
- Protein/peptide name:Z-VEID-FMK
- Purity:>98% (HPLC)
- Formulation:Liquid. In DMSO.
Specifications
Frequently Bought Together