Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Crotalus Atrox 5'-Nucleotidase (From Venom)
Crotalus Atrox 5'-Nucleotidase (From Venom)
Crotalus Atrox 5'-Nucleotidase (From Venom)
Catalog # ENZOBMLKI3075000
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
Crotalus Atrox 5'-Nucleotidase (From Venom)
Catalog # ENZOBMLKI3075000
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pk:
5.000 KU
Source:
Venom
Species:
Crotalus atrox
Seq:
02
Enzyme name:
5'-Nucleotidase
Enzyme activity:
5000 kU
Specifications
About this item
Product Details
Product Details