Human recombinant iNOS,HIS Tag
10 µG
Catalog # ENZOALX2018520010
Supplier: Enzo Life Sciences
Jump to:
Product Details & Documents
Produced in E. coli. Human iNOS is fused to an N-terminal His-tag.
Specifications
Specifications
- Pk:10 µG
- Cat. No.:ENZOALX2018520010
- Protein/peptide type:Recombinant
- Protein/peptide name:iNOS
- Formulation:Liquid. In 50 mM TRIS, pH 7,4, containing 0,1 mM EDTA, 10% glycerol, 100 mM NaCl, 0,1 mM DTT, 10 µM tetrahydrobiopterin.
- Protein synonyms:Inducible NOS|NOS II|Nitric oxide synthase (inducible)
- Purity:>90% (SDS-PAGE)
- Tag sequence:HIS Tag
- Species:Human
- Molecular weight:~130kDa/subunit; homodimer,
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



