Native Hsv-1 antigen
Catalog # ENZOALX6301260200
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:200 µl
- Protein/peptide type:Native
- Seq:02
- Protein/peptide name:Hsv-1 Antigen
- Purity:~80%
- Formulation:Liquid, PBS containing 0,1% Triton X-100
Specifications
About this item
From herpes simplex virus type 1. Inactivated by 0.1% Triton X-100.