Order Entry
Poland
ContactUsLinkComponent
Human recombinant MMP-13
Human recombinant MMP-13
Catalog # ENZOBMLSE4930010
Supplier:  Enzo Life Sciences
CAS Number:  
Human recombinant MMP-13
Catalog # ENZOBMLSE4930010
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    02
  • Protein/peptide name:
    MMP-13

Specifications

About this item

Produced in insect cells. Primarily full length latent MMP-13, with some active forms present.