Order Entry
Poland
ContactUsLinkComponent
Human Recombinant 14-3-3beta (from E. coli), HIS Tag
Human Recombinant 14-3-3beta (from E. coli), HIS Tag
Catalog # ENZOBMLSE4830100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human Recombinant 14-3-3beta (from E. coli), HIS Tag
Catalog # ENZOBMLSE4830100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Tag sequence:
    HIS Tag
  • Protein/peptide name:
    14-3-3 beta
  • Purity:
    >90% (SDS-PAGE)
  • Formulation:
    Liquid, containing 50 mM NaH2PO4, 300 mM NaCl, pH 8,0

Specifications

About this item

Produced in E. coli. Human 14-3-3beta is fused at the C-terminus to a His-tag.