Mouse recombinant netrin 1
Catalog # ENZOALX5221242010
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Species:Mouse
- Seq:02
- Reconstitution instructions:Reconstitute with 100 µl sterile distilled water. Resuspended protein should be used immediately upon reconstitution to avoid loss in activity due to non-specific binding to the vial. Further dilutions should be made with medium containing 5% fetal calf se
- Protein/peptide name:Netrin 1
- Purity:>95% (SDS-PAGE)
- Concentration:0,1 mg/ml after reconstitution.
- Formulation:Lyophilized. Contains PBS.
Specifications
About this item
Produced in HEK 293 cells. The extracellular domain of mouse netrin-1 (aa 27-604) is fused at the C-terminus to a FLAG®-tag.