Human Recombinant CD134L (soluble)
Supplier: Enzo Life Sciences
Certificates
About this item
Produced in HEK 293 cells. The extracellular domain of human CD134L (OX40L) (aa 52-183) is fused at the N-terminus to a linker peptide (8 aa) and a FLAG®-tag.
Specifications
- Protein/peptide type:Recombinant
- Species:Human
- Biological activity:Activates T cell proliferation. Delays neutrophil apoptosis.
- Protein/peptide name:CD134L (soluble)
- Concentration:0,1 mg/ml after reconstitution
- Formulation:Lyophilised. Contains PBS.
- Reconstitution instructions:Reconstitute with 100 µl sterile water. Further dilutions should be made with medium containing 5% fetal calf serum or a carrier protein.
- Purity:>95% (SDS-PAGE)
Specifications
Frequently Bought Together