Human recombinant TRAILR2:Fc, FITC (Fluorescein Isothiocyanate)
Catalog # ENZOALX522005FC050
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:50 µG
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Biological activity:Inhibits human soluble TRAIL-induced apoptosis in a concentration range of 0,5 - 10 µg/ml.
- Protein/peptide name:TRAILR2:Fc
- Purity:>95% (SDS-PAGE)
- Concentration:1 mg/ml
- Formulation:Liquid. In PBS containing 0,02% sodium azide.
Specifications
About this item
Produced in HEK 293 cells. The extracellular domain of human TRAIL-R2 (DR5) (aa 52-212) is fused to the Fc portion of human IgG1.